Anti-PYCR1 Mouse Polyclonal Antibody
Catalog # 103308-836
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:PYCR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:5831
- Antigen Synonyms:PIG45|P5CR|PYCR|proliferation-inducing protein 45|mitochondrial|P5C|PP222|P5CR 1|PRO3|mitochondrial pyrroline-5-carboxylate reductase 1|EC 1.5.1.2|P5C reductase 1|ARCL2B|pyrroline-5-carboxylate reductase 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PYCR1 (NP_008838.2, 1 a.a. - 319 a.a.) full-length human protein. MSVGFIGAGQLAFALAKGFTAAGVLAAHKIMASSPDMDLATVSALRKMGVKLTPHNKETVQHSDVLFLAVKPHIIPFILDEIGADIEDRHIVVSCAAGVTISSIEKKLSAFRPAPRVIRCMTNTPVVVREGATVYATGTHAQVEDGRLMEQLLSSVGFCTEVEEDLIDAVTGLSGSGPAYAFTALDALADGGVKMGLPRRLAVRLGAQALLGAAKMLLHSEQHPGQLKDNVSSPGGATIHALHVLESGGFRSLLINAVEASCIRTRELQSMADQEQVSPAAIKKTILDKVKLDSPAGTALSPSGHTKLLPRSLAPAGKD
- Purification:Protein A purified
- Cat. No.:103308-836
- Supplier no.:H00005831-B01P
Specifications
About this item
The PYCR1 Antibody from Novus Biologicals is a mouse polyclonal antibody to PYCR1. This antibody reacts with human. The PYCR1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: PYCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human