To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:PWP1
- Clonality:Monoclonal
- Clone:3A11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:11137
- Antigen Synonyms:periodic tryptophan protein 1 homolog|Keratinocyte protein IEF SSP 9502|PWP1 homolog (S. cerevisiae)|nuclear phosphoprotein similar to S. cerevisiae PWP1|IEF-SSP-9502
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PWP1 (AAH01652, 1 a.a. ~ 502 a.a) full length recombinant protein with GST tag.MNRSRQVTCVAWVRCGVAKETPDKVELSKEEVKRLIAEAKEK LQEEGGGSDEEETGSPSEDGMQSARTQARPREPLEDGDPEDD RTLDDDELAEYDLDKYDEEGDPDAETLGESLLGLTVYGSNDQ DPYVTLKDTEQYEREDFLIKPSDNLIVCGRAEQDQCNLDVHV YNQEEDSFYVHHDILLSAYPLSVEWLNFDPSPDDSTGNYIAV GNMTPVIEVWDLDIVDSLEPVFTLGSKLSKKKKKK
- Purification:IgG purified
- Cat. No.:103340-216
- Supplier no.:H00011137-M01
Specifications
About this item
The PWP1 Antibody (3A11) from Novus Biologicals is a mouse monoclonal antibody to PWP1. This antibody reacts with human. The PWP1 Antibody (3A11) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: PWP1
Clonality: Monoclonal
Clone: 3A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human