Anti-PLA2G16/HRASLS3 Mouse Polyclonal Antibody
Catalog # 103340-238
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:PLA2G16/HRASLS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:11145
- Antigen Synonyms:group XVI phospholipase A2|Renal carcinoma antigen NY-REN-65|group XVI|phospholipase A2|AdPLAHRASLS3|Adipose-specific phospholipase A2|HREV107-3adipose-specific PLA2|HREV107H-rev 107 protein homolog|HRAS-like suppressor 3Ca-independent phospholipase A1/2|EC 3.1.1.4|MGC118754|H-REV107-1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:HRASLS3 (AAH01387.1, 1 a.a. ~ 162 a.a) full-length human protein.MRAPIPEPKPGDLIEIFRPFYRHWAIYVGDGYVVHLAPPSEVAGAGAASVMSALTDKAIVKKELLYDVAGSDKYQVNNKHDDKYSPLPCSKIIQRAEELVGQEVLYKLTSENCEHFVNELRYGVARSDQVRDVIIAASVAGMGLAAMSLIGVMFSRNKRQKQ
- Purification:Protein A purified
- Cat. No.:103340-238
- Supplier no.:H00011145-B01P
Specifications
About this item
The PLA2G16 / HRASLS3 Antibody from Novus Biologicals is a mouse polyclonal antibody to PLA2G16 / HRASLS3. This antibody reacts with human. The PLA2G16 / HRASLS3 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: PLA2G16/HRASLS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human