Anti-PHYH Mouse Polyclonal Antibody
103307-392
- Antibody Type:Primary
- Antigen Symbol:PHYH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:5264
- Antigen Synonyms:phytanoyl-CoA hydroxylase|LNAP1|PAHXLN1|peroxisomal|phytanoyl-CoA 2-hydroxylase|phytanoyl-CoA hydroxylase (Refsum disease)|PHYH1|RD|PhyH
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PHYH (NP_006205.1, 1 a.a. - 338 a.a.) full-length human protein. MEQLRAAARLQIVLGHLGRPSAGAVVAHPTSGTISSASFHPQQFQYTLDNNVLTLEQRKFYEENGFLVIKNLVPDADIQRFRNEFEKICRKEVKPLGLTVMRDVTISKSEYAPSEKMITKVQDFQEDKELFRYCTLPEILKYVECFTGPNIMAMHTMLINKPPDSGKKTSRHPLHQDLHYFPFRPSDLIVCAWTAMEHISRNNGCLVVLPGTHKGSLKPHDYPKWEGGVNKMFHGIQDYEENKARVHLVMEKGDTVFFHPLLIHGSGQNKTQGFRKAISCHFASADCHYIDVKGTSQENIEKEVVGIAHKFFGAENSVNLKDIWMFRARLVKGERTNL
- Purification:Protein A purified
- Cat. No.:103307-392
- Supplier no.:H00005264-B01P
The PHYH Antibody from Novus Biologicals is a mouse polyclonal antibody to PHYH. This antibody reacts with human. The PHYH Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: PHYH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human