Anti-Pax2 Mouse Monoclonal Antibody [clone: 3C7]
Catalog # 103332-270
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:Pax2
- Clonality:Monoclonal
- Clone:3C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:5076
- Antigen Synonyms:paired box homeotic gene 2|paired box 2|paired box protein Pax-2|paired box gene 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PAX2 (NP_000269 194 a.a. - 303 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PRSNGEKRKRDEDVSEGSVPNGDSQSGVDSLRKHLRADTFTQQQLEALDRVFERPSYPDVFQASEHIKSEQGNEYSLPALTPGLDEVKSSLSASTNPELGSNVSGTQTYP
- Purification:IgG purified
- Cat. No.:103332-270
- Supplier no.:H00005076-M01
Specifications
About this item
The Pax2 Antibody (3C7) from Novus Biologicals is a mouse monoclonal antibody to Pax2. This antibody reacts with human. The Pax2 Antibody (3C7) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Pax2
Clonality: Monoclonal
Clone: 3C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human