Anti-p27/Kip1 Mouse Monoclonal Antibody [clone: SPM348]
Catalog # 103287-950
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:p27/Kip1
- Clonality:Monoclonal
- Clone:SPM348
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Primates,Human,Rat,Mouse
- Size:7 ml
- Gene ID:1027
- Antigen Synonyms:Cyclin-dependent kinase inhibitor p27|CDKN4|cyclin-dependent kinase inhibitor 1B (p27|MEN1B|Kip1)|MEN4|cyclin-dependent kinase inhibitor 1B|p27Kip1|KIP1P27KIP1
- Storage Buffer:10mM PBS and 0.05% BSA
- Storage Temperature:Store at 4C.
- Immunogen:Purified GST-p27 fusion protein of human origin
- Purification:Protein A or G purified
- Cat. No.:103287-950
- Supplier no.:NBP2-44494
Specifications
About this item
The p27 / Kip1 Antibody (SPM348) - IHC-Prediluted from Novus Biologicals is a mouse monoclonal antibody to p27 / Kip1. This antibody reacts with human, mouse, rat, primate. The p27 / Kip1 Antibody (SPM348) - IHC-Prediluted has been validated for the following applications: Immunohistochemistry-Paraffin.
Type: Primary
Antigen: p27/Kip1
Clonality: Monoclonal
Clone: SPM348
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Primate