Anti-NUDCD3 Mouse Polyclonal Antibody
103341-264
- Antibody Type:Primary
- Antigen Symbol:NUDCD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:23386
- Antigen Synonyms:KIAA1068NudCL|NudC domain containing 3|NudC-like protein|nudC domain-containing protein 3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:NUDCD3 (AAH03691.1, 1 a.a. ~ 220 a.a) full-length human protein.MAHGSQEAEAPGAVAGAAEVPREPPILPRIQEQFQKNPDSYNGAVRENYTWSQDYTDLEVRVPVPKHVVKGKQVSVALSSSSIRVAMLEENGERVLMEGKLTHKINTESSLWSLEPGKCVLVNLSKVGEYWWNAILEGEEPIDIDKINKERSMATVDEEEQAVLDRLTFDYHQKLQGKPQSHELKVHEMLKKGWDAEGSPFRGQRFDPAMFNISPGAVQF
- Purification:Protein A purified
- Cat. No.:103341-264
- Supplier no.:H00023386-B01P
The NUDCD3 Antibody from Novus Biologicals is a mouse polyclonal antibody to NUDCD3. This antibody reacts with human. The NUDCD3 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: NUDCD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human