To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:NFkB p105/p50
- Clonality:Monoclonal
- Clone:2J10D7
- Conjugation:Biotin
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:4790
- Antigen Synonyms:DKFZp686C01211|EBP-1|p50|nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151|NFKB-p50|NFKB-p105|nuclear factor kappa-B DNA binding subunit|nuclear factor NF-kappa-B p105 subunit|NF-kappaB|DNA-binding factor KBF1|nuclear factor NF-kappa-B p50 subunit|p105|KBF1|NF-kappabeta|NF-kappa-B|DNA binding factor KBF1
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A portion of amino acids 150-200 of human NF-kB (p50) was used as the immunogen.
- Purification:Protein G purified
- Cat. No.:103360-480
The NFkB p105 / p50 Antibody (2J10D7) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to NFkB p105 / p50. This antibody reacts with human. The NFkB p105 / p50 Antibody (2J10D7) [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: NFkB p105/p50
Clonality: Monoclonal
Clone: 2J10D7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human