MycoClean Mycoplasma Prevention Spray, Bulldog Bio
Catalog # 103507-506
Supplier: Bulldog Bio
Specifications
- Description:Mycoplasma Prevention Spray
- Size:200 mL
- Cat. No.:103507-506
Specifications
About this item
MycoClean Mycoplasma Prevention Spray is the lab's solution for decontamination of the lab's cell culture areas, incubators, work benches, storage containers and apparatuses.
- Ready-to-use
- Non-corrosive and non-carcinogenic
- Can be stored at room temperature for two years
Mycoplasma lacks a cell wall, making them unresponsive to most antibiotic treatments
Prevention spray forms pores in cell membrane using osmotic pressure and eradicate mycoplasma.
With this easy and convenient spray, contamination-free surfaces can be achieved with a single application.
Caution: For in vitro research only. Not for human diagnostic or therapeutics.