Multistix® Urinalysis Reagent Test Strips, Siemens Healthineers
Supplier: Siemens Healthineers
Certificates
About this item
Multistix® Urinalysis Reagent Test Strips from Siemens Healthineers are the reliable frontline test for detection of a broad range of conditions, from detecting urinary tract infections (UTI1) to diabetes and kidney disorders.
- Reliable testing
- Detects a range of conditions
- Fewer false negatives
Accurately gain a broader clinical insight into a patient's health. Reliable testing for UTI, diabetes, and kidney disorders. 10 tests for important markers on a single strip. Combined leukocyte/nitrite results for better prediction of UTI with fewer false negatives. 100 strips per bottle.