ContactUsLinkComponent
CLINITEK® Microalbumin Urinalysis Reagent Test Strips, Siemens Healthineers
CLINITEK® Microalbumin Urinalysis Reagent Test Strips, Siemens Healthineers
  89140-216
 :  
undefined
CLINITEK® Microalbumin Urinalysis Reagent Test Strips, Siemens Healthineers
  89140-216
 :  10317439
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    CLINITEK Microalbumin 2 Urinalysis Reagent Test Strips
  • Cat. No.:
    89140-216
  • Parameters tested:
    Microalbumin, Creatinine
  • Supplier no.:
    2083
CLINITEK® Microalbumin 2 Urinalysis Reagent Test Strips from Siemens Healthcare Diagnostics provide albumin, creatinine and albumin-to-creatinine ratio results in one minute.
  • Results in one minute
  • Early kidney disease detection
  • Increased sensitivity of albumin
The CLINITEK® Microalbumin strips are useful to test for microalbuminuria in patients with diabetes or hypertension in order to detect early kidney disease
CLINITEK® Microalbumin Urinalysis Reagent Test Strips make it simple and easy to test patients with diabetes. Because testing for microalbuminuria can detect kidney disease in its early stages, physicians may begin intervention earlier than might otherwise have been possible. These strips permit detection of elevated albumin sooner, more sensitively and more specifically than products designed for general protein testing. These strips also provide semi-quantitative results with a hard-copy report on the CLINITEK Status® family of analyzers, making them ideal for point-of-care testing. Patient samples can be tested immediately during a physician, clinic or hospital visit, rather than sending them out and waiting for results. The strips are also convenient for testing random urine samples, eliminating the need to collect a timed sample. The CLINITEK Microalbumin Urinalysis Reagent Test Strips are CLIA-waived for use on the CLINITEK 50® and CLINITEK Status Family of Analyzers.
The strips are used on the CLINITEK Status® family of analyzers. Albumin, creatinine, and albumin-to-creatinine ratio results to help identify clinically relevant microalbuminuria. Increased sensitivity of albumin as recommended by ADA guidelines: sensitivity as low as 2.0 mg/dL. Results are corrected for varying urine concentrations when used on the CLINITEK Status family of analyzers, replacing the need for 24-hour urine collection and measurement.