Powder pillows, CyaniVer® reagents for cyanide determination, Hach
Supplier: Avantor
Certificates
About this item
For cyanide determination by the pyridine-pyrazolone method (Hach method 8027).
Packaging: Pack of 100 powder pillows.
Specifications
- Method:8027 - Pyridine-Pyrazalone