Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Mohawk homeobox
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:283078
- Antigen Synonyms:iroquois homeobox protein-like 1|Iroquois family related homeodomain protein|mohawk homeobox|homeobox protein Mohawk|MGC39616|C10orf48|IFRX|IRXL1chromosome 10 open reading frame 48
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:0.7 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human MKX. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102709-628
- Supplier no.:NBP2-19378
Specifications
About this item
The Mohawk homeobox Antibody from Novus Biologicals is a rabbit polyclonal antibody to Mohawk homeobox. This antibody reacts with human. The Mohawk homeobox Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Mohawk homeobox
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human