Anti-Mitochondrial-processing peptidase subunit beta Mouse Monoclonal Antibody [clone: 3C3]
Catalog # 103337-130
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Mitochondrial-processing peptidase subunit beta
- Clonality:Monoclonal
- Clone:3C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:9512
- Antigen Synonyms:Beta-MPP|EC 3.4.24.64|P-52|peptidase (mitochondrial processing) beta|MPPBMPP11|MPPP52|mitochondrial processing peptidase beta subunit|mitochondrial-processing peptidase subunit beta
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:PMPCB (NP_004270, 161 a.a. ~ 270 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.IQNSTLGEAEIERERGVILREMQEVETNLQEVVFDYLHATAYQNTALGRTILGPTENIKSISRKDLVDYITTHYKGPRIVLAAAGGVSHDELLDLAKFHFGDSLCTHKG
- Purification:Protein A purified
- Cat. No.:103337-130
- Supplier no.:H00009512-M01
Specifications
About this item
The Mitochondrial-processing peptidase subunit beta Antibody (3C3) from Novus Biologicals is a mouse monoclonal antibody to Mitochondrial-processing peptidase subunit beta. This antibody reacts with human. The Mitochondrial-processing peptidase subunit beta Antibody (3C3) has been validated for the following applications: ELISA.
Type: Primary
Antigen: Mitochondrial-processing peptidase subunit beta
Clonality: Monoclonal
Clone: 3C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human