Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Methionyl tRNA synthetase 2 Mouse Polyclonal Antibody
Anti-Methionyl tRNA synthetase 2 Mouse Polyclonal Antibody
Catalog # 103345-922
Supplier:  Novus Biologicals
Anti-Methionyl tRNA synthetase 2 Mouse Polyclonal Antibody
Catalog # 103345-922
Supplier:  Novus Biologicals
Supplier Number:  H00092935-B01P

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Methionyl tRNA synthetase 2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.05 mg
  • Gene ID:
    92935
  • Antigen Synonyms:
    Mitochondrial methionine--tRNA ligase|mitochondrial methionyl-tRNA synthetase|EC 6.1.1.10|methionine tRNA ligase 2|MetRS|mitochondrial|methionine-tRNA synthetase 2 (mitochondrial)|methionyl-tRNA synthetase 2|methionine-tRNA synthetase 2|methionine-tRNA ligase|methionyl-tRNA synthetase|EC 6.1.1|Methionine--tRNA ligase 2|mtMetRS
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    MARS2 (NP_612404.1, 1 a.a. ~ 593 a.a) full-length human protein.
  • Purification:
    Protein G purified
  • Cat. No.:
    103345-922
  • Supplier no.:
    H00092935-B01P

Specifications

About this item

The Methionyl tRNA synthetase 2 Antibody from Novus Biologicals is a mouse polyclonal antibody to Methionyl tRNA synthetase 2. This antibody reacts with human. The Methionyl tRNA synthetase 2 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Methionyl tRNA synthetase 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human