Anti-Integrin alpha V beta 3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 23C6]
Catalog # 103357-240
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Integrin alpha V beta 3
- Clonality:Monoclonal
- Clone:23C6
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μl
- Gene ID:3685
- Antigen Synonyms:alpha V (vitronectin receptor|MSK8|Vitronectin receptor subunit alpha|CD51|VNRADKFZp686A08142|antigen identified by monoclonal L230|alpha polypeptide|CD51 antigen|antigen CD51)|integrin alpha-V|integrin
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A cell suspension containing osteoclasts from osteoclastomas.
- Purification:Protein A or G purified
- Cat. No.:103357-240
Specifications
About this item
The Integrin alpha V beta 3 Antibody (23C6) [PE] from Novus Biologicals is a mouse monoclonal antibody to Integrin alpha V beta 3. This antibody reacts with human. The Integrin alpha V beta 3 Antibody (23C6) [PE] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: Integrin alpha V beta 3
Clonality: Monoclonal
Clone: 23C6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human