To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Glycophorin C
- Clonality:Monoclonal
- Clone:BRIC4
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2995
- Antigen Synonyms:PAS-2|MGC117309|glycophorin-D|Glycoprotein beta|CD236 antigen|Sialoglycoprotein D|CD236|PAS-2'|glycophorin-C|GPD|glycoconnectin|CD236R|glycophorin C (Gerbich blood group)|MGC126192|GYPD|GLPC|GE|GPCMGC126191
- Storage Buffer:TBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Human erythrocytes
- Tested Applications:Haemagglutination
- Purification:Protein A or G purified
- Cat. No.:103282-866
- Supplier no.:NBP2-33304
Specifications
About this item
The Glycophorin C Antibody (BRIC4) from Novus Biologicals is a mouse monoclonal antibody to Glycophorin C. This antibody reacts with human. The Glycophorin C Antibody (BRIC4) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Haemagglutination.
Type: Primary
Antigen: Glycophorin C
Clonality: Monoclonal
Clone: BRIC4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human