Anti-NAV1 Rabbit Polyclonal Antibody
103259-472
- Antibody Type:Primary
- Antigen Symbol:NAV1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:89796
- Antigen Synonyms:unc53H1|pore membrane and/or filament interacting like protein 3|MGC14961|Steerin-1|neuron navigator 1|KIAA1213|FLJ14203|KIAA1151POMFIL3DKFZp781D0314|STEERIN1|neuron navigator-1|FLJ12560|Pore membrane and/or filament-interacting-like protein 3|Unc-53 homolog 1
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:AKSFVKPPSLANLDKVNSNSLDLPSSSDTTHASKVPDLHATSSASGGPLPSCFTPSPAPILNINSASFSQGLELMSGFSVPKETRMYPKLSGLHRSMESLQMPMSLPSAFPSSTPVPTPPAPPAAPTEEETEELTWSGSP
- Purification:Immunogen affinity purified
- Cat. No.:103259-472
- Supplier no.:NBP1-84082
The NAV1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NAV1. This antibody reacts with human. The NAV1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: NAV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human