Anti-MSH2 Rabbit Polyclonal Antibody
Catalog # 103359-094
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:MSH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.025 mg
- Gene ID:4436
- Antigen Synonyms:DNA mismatch repair protein Msh2|COCA1|MutS protein homolog 2|colon cancer|nonpolyposis type 1 (E. coli)|nonpolyposis type 1)|FCC1|HNPCC|HNPCC1mutS (E. coli) homolog 2 (colon cancer|hMSH2|mutS homolog 2|LCFS2
- Storage Buffer:PBS containing 0.2% gelatin
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:A portion of amino acids 550-600 of human MSH2 was used as the immunogen.
- Purification:Protein G purified
- Cat. No.:103359-094
Specifications
About this item
The MSH2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to MSH2. This antibody reacts with human, mouse. The MSH2 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: MSH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse