Anti-MPP1 Mouse Monoclonal Antibody [clone: 1C3-1D11]
103331-034
- Antibody Type:Primary
- Antigen Symbol:MPP1
- Clonality:Monoclonal
- Clone:1C3-1D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:4354
- Antigen Synonyms:AAG12|palmitoylated erythrocyte membrane protein|PEMPpalmitoylated 1 (55kD)|Membrane protein|MRG1|EC 2.7.11|55 kDa erythrocyte membrane protein|palmitoylated 1|55kDa|membrane protein
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MPP1 (AAH02392, 1 a.a. - 466 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTLKASEGESGGSMHTALSDLYLEHLLQKRSRPEAVSHPLNTVTEDMYTNGSPAPGSPAQVKGQEVRKVRLIQFEKVTEEPMGITLKLNEKQSCTVARILHGGMIHRQGSLHVGDEILEINGTNVTNHSVDQLQKAMKETKGMISLKVIPNQQSRLPALQMFMRAQFDYDPKKDNLIPCKEAGLKFATGDIIQIINKDDSNWWQGRVEGSSKESAGLIPSPELQEWRVASMAQSAPSEAPSCSPFGKKKKYKDKYLAKHSSIFDQLDVVSYEEVVRLPAFKRKTLVLIGASGVGRSHIKNALLSQNPEKFVYPVPYTTRPPRKSEEDGKEYHFISTEEMTRNISANEFLEFGSYQGNMFGTKFETVHQIHKQNKIAILDIEPQTLKIVRTAELSPFIVFIAPTDQGTQTEALQQLQKDSEAIRSQYAHYFDLSLVNNGVDETLKKLQEAFDQACSSPQWVPVSWVY
- Tested Applications:RNA Inhibition
- Purification:IgG purified
- Cat. No.:103331-034
- Supplier no.:H00004354-M02
The MPP1 Antibody (1C3-1D11) from Novus Biologicals is a mouse monoclonal antibody to MPP1. This antibody reacts with human. The MPP1 Antibody (1C3-1D11) has been validated for the following applications: Western Blot, ELISA, RNA Inhibition.
Type: Primary
Antigen: MPP1
Clonality: Monoclonal
Clone: 1C3-1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human