Anti-MKP-3/DUSP6 Goat Polyclonal Antibody
103408-814
- Antibody Type:Primary
- Antigen Symbol:MKP-3/DUSP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:1848
- Antigen Synonyms:MKP3serine/threonine specific protein phosphatase|Dual specificity protein phosphatase PYST1|PYST1dual specificity protein phosphatase 6|MKP-3|dual specificity phosphatase 6|MAP kinase phosphatase 3|EC 3.1.3.16|Mitogen-activated protein kinase phosphatase 3|EC 3.1.3.48
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-PSNQNVYQVDSLQST corresponding to C-Terminus according to NP_073143.2.
- Tested Applications:Peptide
- Purification:Immunogen affinity purified
- Cat. No.:103408-814
The MKP-3 / DUSP6 Antibody from Novus Biologicals is a goat polyclonal antibody to MKP-3 / DUSP6. This antibody reacts with human, mouse, rat, porcine. The MKP-3 / DUSP6 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Type: Primary
Antigen: MKP-3/DUSP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine