Anti-MDH2 Mouse Monoclonal Antibody [clone: 2D9]
103330-654
- Antibody Type:Primary
- Antigen Symbol:MDH2
- Clonality:Monoclonal
- Clone:2D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:4191
- Antigen Synonyms:NAD (mitochondrial)|MGC:3559|mitochondrial|EC 1.1.1.37|malate dehydrogenase 2|EC 1.1.1|MOR1|MDH
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MDH2 (NP_005909 134 a.a. - 246 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EAMICVIANPVNSTIPITAEVFKKHGVYNPNKIFGVTTLDIVRANTFVAELKGLDPARVNVPVIGGHAGKTIIPLISQCTPKVDFPQDQLTALTGRIQEAGTEVVKAKAGAGS
- Purification:IgG purified
- Cat. No.:103330-654
- Supplier no.:H00004191-M02
The MDH2 Antibody (2D9) from Novus Biologicals is a mouse monoclonal antibody to MDH2. This antibody reacts with human. The MDH2 Antibody (2D9) has been validated for the following applications: ELISA.
Type: Primary
Antigen: MDH2
Clonality: Monoclonal
Clone: 2D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human