Anti-MCAT Rabbit Polyclonal Antibody
Catalog # 103342-832
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:MCAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:27349
- Antigen Synonyms:FASN2C|fabD|MTMGC47838|MCT[Acyl-carrier-protein] malonyltransferase|mitochondrial|EC 2.3.1.39|malonyl-CoA-acyl carrier protein transacylase|malonyl-CoA:acyl carrier protein transacylase|NET62|malonyl CoA:ACP acyltransferase (mitochondrial)|Mitochondrial malonyltransferase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MCAT (NP_055322.1, 1 a.a. ~ 180 a.a) full-length human protein.MSVRVARVAWVRGLGASYRRGASSFPVPPPGAQGVAELLRDATGAEEEAPWAATERRMPGQCSVLLFPGQGSQVVGMGRGLLNYPRVRELYAAARRVLGYDLLELSLHGPQETLDRTVHCQPAIFVASLAAVEKLHHLQPSVIENCVAAAGFSVGEFAALVFAGAMEFAEGSTVSPEEFL
- Purification:Protein A purified
- Cat. No.:103342-832
- Supplier no.:H00027349-D01P
Specifications
About this item
The MCAT Antibody from Novus Biologicals is a rabbit polyclonal antibody to MCAT. This antibody reacts with human. The MCAT Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: MCAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human