Anti-MAGI2 Mouse Monoclonal Antibody [clone: 6C8]
103337-714
- Antibody Type:Primary
- Antigen Symbol:MAGI2
- Clonality:Monoclonal
- Clone:6C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:9863
- Antigen Synonyms:atrophin-1 interacting protein A|WW and PDZ domain-containing protein 2|KIAA0705activin receptor interacting protein 1|membrane-associated guanylate kinase|WW and PDZ domain containing 2|atrophin 1 interacting protein 1|AIP1AIP-1|Atrophin-1-interacting protein 1|Membrane-associated guanylate kinase inverted 2|membrane associated guanylate kinase|Atrophin-1-interacting protein A|MAGI-2SSCAM|ARIP1|ACVRINP1|ACVRIP1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MAGI2 (NP_036433, 519 a.a. ~ 629 a.a) partial recombinant protein with GST tag.PANSMVPPLAIMERPPPVMVNGRHNYETYLEYISRTSQSVPDITDRPPHSLHSMPTDGQLDGTYPPPVHDDNVSMASSGATQAELMTLTIVKGAQGFGFTIADSPTGQRV
- Purification:IgG purified
- Cat. No.:103337-714
- Supplier no.:H00009863-M01
The MAGI2 Antibody (6C8) from Novus Biologicals is a mouse monoclonal antibody to MAGI2. This antibody reacts with human. The MAGI2 Antibody (6C8) has been validated for the following applications: ELISA.
Type: Primary
Antigen: MAGI2
Clonality: Monoclonal
Clone: 6C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human