Anti-LSD1 Rabbit Polyclonal Antibody
Catalog # 103359-214
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:LSD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Zebrafish,Primates,Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:0.025 mg
- Gene ID:23028
- Antigen Synonyms:BHC110FAD-binding protein BRAF35-HDAC complex|Flavin-containing amine oxidase domain-containing protein 2|110 kDa subunit|AOF2lysine-specific histone demethylase 1|amine oxidase (flavin containing) domain 2|lysine (K)-specific demethylase 1|KIAA0601KDM1|lysine (K)-specific demethylase 1A|lysine-specific histone demethylase 1A|LSD1BRAF35-HDAC complex protein BHC110
- Storage Buffer:PBS containing 0.2% gelatin
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:A portion of amino acids 100-150 of human LSD1 was used as the immunogen.
- Purification:Immunogen affinity purified
- Cat. No.:103359-214
Specifications
About this item
The LSD1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to LSD1. This antibody reacts with human, mouse, rat, canine, primate, zebrafish. The LSD1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: LSD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Canine, Primate, Zebrafish