To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:JNK3
- Clonality:Monoclonal
- Clone:3A3-1A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:5602
- Antigen Synonyms:JNK3A|MGC50974|JNK3 alpha protein kinase|PRKM10FLJ33785|MAP kinase 10|MAP kinase p49 3F12|Stress-activated protein kinase JNK3|p493F12|mitogen-activated protein kinase 10|stress activated protein kinase beta|EC 2.7.11|p54bSAPK|c-Jun N-terminal kinase 3|JNK3FLJ12099|MAPK 10|EC 2.7.11.24
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MAPK10 (AAH65516.1, 1 a.a. - 319 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MELMDANLCQVIQMELDHERMSYLLYQMLCGIKHLHSAGIIHRDLKPSNIVVKSDCTLKILDFGLARTAGTSFMMTPYVVTRYYRAPEVILGMGYKENVDIWSVGCIMGEMVRHKILFPGRDYIDQWNKVIEQLGTPCPEFMKKLQPTVRNYVENRPKYAGLTFPKLFPDSLFPADSEHNKLKASQARDLLSKMLVIDPAKRISVDDALQHPYINVWYDPAEVEAPPPQIYDKQLDEREHTIEEWKELIYKEVMNSEEKTKNGVVKGQPSPSGAAVNSSESLPPSSSVNDISSMSTDQTLASDTDSSLEASAGPLGCCR
- Purification:IgG purified
- Cat. No.:103308-222
- Supplier no.:H00005602-M03
Specifications
About this item
The JNK3 Antibody (3A3-1A8) from Novus Biologicals is a mouse monoclonal antibody to JNK3. This antibody reacts with human. The JNK3 Antibody (3A3-1A8) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: JNK3
Clonality: Monoclonal
Clone: 3A3-1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human