Anti-IL-27 Mouse Monoclonal Antibody [clone: 3F12]
Catalog # 103347-906
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Symbol:IL-27
- Clonality:Monoclonal
- Clone:3F12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:246778
- Antigen Synonyms:IL-27 subunit alpha|IL-27-A|IL27p28|IL27A|interleukin 30|IL-27|p28IL-27A|interleukin-27 subunit alpha|IL-27 p28 subunit|IL30|IL27-A|interleukin 27|MGC71873
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:IL27 (NP_663634, 177 a.a. ~ 244 a.a) partial recombinant protein with GST tag.RKGLLPGALGSALQGPAQVSWPQLLSTYRLLHSLELVLSRAV RELLLLSKAGHSVWPLGFPTLSPQP
- Purification:IgG purified
- Cat. No.:103347-906
- Supplier no.:H00246778-M01
Specifications
About this item
The IL-27 Antibody (3F12) from Novus Biologicals is a mouse monoclonal antibody to IL-27. This antibody reacts with human. The IL-27 Antibody (3F12) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.
Type: Primary
Antigen: IL-27
Clonality: Monoclonal
Clone: 3F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human