Anti-IL-17 RA/IL-17 R Mouse Monoclonal Antibody [clone: 49M4D2]
Catalog # 103487-740
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:IL-17 RA/IL-17 R
- Clonality:Monoclonal
- Clone:49M4D2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:23765
- Antigen Synonyms:IL17Rinterleukin-17 receptor A|CD217 antigen|interleukin 17 receptor A|IL-17 receptor A|IL-17RAMGC10262|CD217|hIL-17R|CDw217interleukin 17 receptor
- Storage Buffer:0.1 ml PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Within the range of amino acids 186-267 of human IL-17RA were used as the immunogen for the antibody.
- Purification:Protein G purified
- Cat. No.:103487-740
Specifications
About this item
The IL-17 RA / IL-17 R Antibody (49M4D2) from Novus Biologicals is a mouse monoclonal antibody to IL-17 RA / IL-17 R. This antibody reacts with human. The IL-17 RA / IL-17 R Antibody (49M4D2) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: IL-17 RA/IL-17 R
Clonality: Monoclonal
Clone: 49M4D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human