Anti-IKK beta Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10AG2]
103359-442
- Antibody Type:Primary
- Antigen Symbol:IKK beta
- Clonality:Monoclonal
- Clone:10AG2
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Size:100 μl
- Gene ID:3551
- Antigen Synonyms:MGC131801|I-kappa-B kinase 2|IKKBFLJ40509|IKK-B|NFKBIKBFLJ33771|IkBKB|inhibitor of kappa light polypeptide gene enhancer in B-cells|kinase beta|I-kappa-B-kinase beta|Nuclear factor NF-kappa-B inhibitor kinase beta|EC 2.7.11|IKK-betaFLJ36218|IKK2FLJ38368|EC 2.7.11.10|inhibitor of nuclear factor kappa-B kinase subunit beta
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A full-length human IKKb recombinant protein was used to immunize mice (NP_001547).
- Purification:Protein G purified
- Cat. No.:103359-442
The IKK beta Antibody (10AG2) [PE] from Novus Biologicals is a mouse monoclonal antibody to IKK beta. This antibody reacts with human, mouse. The IKK beta Antibody (10AG2) [PE] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: IKK beta
Clonality: Monoclonal
Clone: 10AG2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse