Anti-IkB-alpha Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6A920]
Catalog # 103359-390
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:IkB-alpha
- Clonality:Monoclonal
- Clone:6A920
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Size:100 μl
- Gene ID:4792
- Antigen Synonyms:MAD3|alpha|IkappaBalphaNF-kappa-B inhibitor alpha|nuclear factor of kappa light chain gene enhancer in B-cells|IkB-alpha|IKBAikB-alpha|Major histocompatibility complex enhancer-binding protein MAD3|NFKBI|nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor|I-kappa-B-alpha|MAD-3
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A recombinant protein corresponding to amino acid residues 32-291 of human IkBa was used as immunogen.
- Purification:Protein G purified
- Cat. No.:103359-390
Specifications
About this item
The IkB-alpha Antibody (6A920) [PE] from Novus Biologicals is a mouse monoclonal antibody to IkB-alpha. This antibody reacts with human, mouse, rat. The IkB-alpha Antibody (6A920) [PE] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: IkB-alpha
Clonality: Monoclonal
Clone: 6A920
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat