Anti-IGF2BP3 Rabbit Polyclonal Antibody
Catalog # 103260-264
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:IGF2BP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:10643
- Antigen Synonyms:KH domain containing protein overexpressed in cancer|CT98|IMP3VICKZ family member 3|IGF2 mRNA-binding protein 3|hKOC|IGF-II mRNA-binding protein 3|KOC1DKFZp686F1078|IGF II mRNA binding protein 3|IMP-3KH domain-containing protein overexpressed in cancer|VICKZ3cancer/testis antigen 98|insulin-like growth factor 2 mRNA binding protein 3|insulin-like growth factor 2 mRNA-binding protein 3
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:VVESCEQVNTDSETAVVNVTYSSKDQARQALDKLNGFQLENFTLKVAYIPDEMAAQQNPLQQPRGRRGLGQRGSSRQGSPGSVSKQKPCDLPLRLLVPTQFVGAIIGKEGATIRNITKQTQ
- Purification:Immunogen affinity purified
- Cat. No.:103260-264
- Supplier no.:NBP1-84339
Specifications
About this item
The IGF2BP3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to IGF2BP3. This antibody reacts with human, mouse, rat. The IGF2BP3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: IGF2BP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat