Anti-IFNA2 Mouse Monoclonal Antibody [clone: 1C8]
Catalog # 103328-860
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Interferon 2
- Antigen Symbol:IFNA2
- Clonality:Monoclonal
- Clone:1C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:3440
- Antigen Synonyms:LeIF A|interferon|MGC125764|IFN-alphaA|alpha-2a interferon|alpha 2|INFA2|IFN-alpha-2|interferon alpha 2b|interferon alpha A|MGC125765|IFNA|Interferon alpha-A|interferon alpha-2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:IFNA2 (NP_000596, 24 a.a. - 188 a.a.) partial recombinant protein. MDLPQTHSLGSRRTLMLLAQMRRISLFSCLKDRHDFGFPQEEFGNQFQKAETIPVLHEMIQQIFNLFSTKDSSAAWDETLLDKFYTELYQQLNDLEACVIQGVGVTETPLMKEDSILAVRKYFQRITLYLKEKKYSPCAWEVVRAEIMRSFSLSTNLQESLRSKE
- Purification:IgG purified
- Cat. No.:103328-860
- Supplier no.:H00003440-M07
Specifications
About this item
The IFN-alpha 2 Antibody (1C8) from Novus Biologicals is a mouse monoclonal antibody to IFN-alpha 2. This antibody reacts with human. The IFN-alpha 2 Antibody (1C8) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: IFNA2
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human