Anti-HIF-1 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 647)
103368-148
- Antibody Type:Primary
- Antigen Symbol:HIF-1 alpha
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Zebrafish,Primates,Xenopus,Human,Guinea pig,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:3091
- Antigen Synonyms:HIF1A|hypoxia inducible factor 1|HIF1-alpha|member of PAS superfamily 1|PAS domain-containing protein 8|PASD8alpha subunit (basic helix-loop-helix transcriptionfactor)|BHLHE78|ARNT-interacting protein|Basic-helix-loop-helix-PAS protein MOP1|hypoxia-inducible factor 1-alpha|MOP1HIF1-ALPHA|HIF-1-alpha|HIF-1 alpha|alpha subunit (basic helix-loop-helix transcriptionfactor)|Member of PAS protein 1|Class E basic helix-loop-helix protein 78
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A fusion protein made to an internal sequence of human HIF-1 alpha (containing amino acids 432 - 528) [UniProt Q16665].
- Purification:Immunogen affinity purified
- Cat. No.:103368-148
The HIF-1 alpha Antibody [Alexa Fluor« 647] from Novus Biologicals is a rabbit polyclonal antibody to HIF-1 alpha. This antibody reacts with human, mouse, rat, canine, guinea pig, primate, xenopus, zebrafish. The HIF-1 alpha Antibody [Alexa Fluor« 647] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: HIF-1 alpha
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Canine, Guinea Pig, Primate, Xenopus, Zebrafish