To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:HIF-1 alpha
- Clonality:Monoclonal
- Clone:H1alpha 67-7
- Conjugation:Alexa Fluor® 700
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Monkey,Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:3091
- Antigen Synonyms:HIF1A|hypoxia inducible factor 1|HIF1-alpha|member of PAS superfamily 1|PAS domain-containing protein 8|PASD8alpha subunit (basic helix-loop-helix transcriptionfactor)|BHLHE78|ARNT-interacting protein|Basic-helix-loop-helix-PAS protein MOP1|hypoxia-inducible factor 1-alpha|MOP1HIF1-ALPHA|HIF-1-alpha|HIF-1 alpha|alpha subunit (basic helix-loop-helix transcriptionfactor)|Member of PAS protein 1|Class E basic helix-loop-helix protein 78
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A fusion protein containing amino acids 432 - 528 of human HIF-1 alpha [UniProt# Q16665].
- Purification:Protein G purified
- Cat. No.:103368-074
Specifications
About this item
The HIF-1 alpha Antibody (H1alpha 67-7) [Alexa Fluor« 700] from Novus Biologicals is a mouse monoclonal antibody to HIF-1 alpha. This antibody reacts with human, monkey. The HIF-1 alpha Antibody (H1alpha 67-7) [Alexa Fluor« 700] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: HIF-1 alpha
Clonality: Monoclonal
Clone: H1alpha 67-7
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey