Anti-HIF-1 alpha Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: H1alpha67]
103367-854
- Antibody Type:Primary
- Antigen Symbol:HIF-1 alpha
- Clonality:Monoclonal
- Clone:H1alpha67
- Conjugation:Alexa Fluor® 700
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Yeast,Mouse,Primates,Pig,Rabbit,Monkey,Xenopus,Sheep,Ferret,Human,Bovine,Rat
- Western Blot:Yes
- Size:100 μl
- Gene ID:3091
- Antigen Synonyms:HIF1A|hypoxia inducible factor 1|HIF1-alpha|member of PAS superfamily 1|PAS domain-containing protein 8|PASD8alpha subunit (basic helix-loop-helix transcriptionfactor)|BHLHE78|ARNT-interacting protein|Basic-helix-loop-helix-PAS protein MOP1|hypoxia-inducible factor 1-alpha|MOP1HIF1-ALPHA|HIF-1-alpha|HIF-1 alpha|alpha subunit (basic helix-loop-helix transcriptionfactor)|Member of PAS protein 1|Class E basic helix-loop-helix protein 78
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A fusion protein containing amino acids 432 - 528 of human HIF-1 alpha [UniProt# Q16665].
- Purification:Protein G purified
- Cat. No.:103367-854
The HIF-1 alpha Antibody (H1alpha67) [Alexa Fluor« 700] from Novus Biologicals is a mouse monoclonal antibody to HIF-1 alpha. This antibody reacts with human, mouse, rat, porcine, bovine, ferret, monkey, primate, rabbit, sheep, xenopus, yeast. The HIF-1 alpha Antibody (H1alpha67) [Alexa Fluor« 700] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: HIF-1 alpha
Clonality: Monoclonal
Clone: H1alpha67
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Ferret, Monkey, Primate, Rabbit, Sheep, Xenopus, Yeast