Anti-GPD1L Rabbit Polyclonal Antibody
Catalog # 103341-044
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:GPD1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:23171
- Antigen Synonyms:EC 1.1.1.8|glycerol-3-phosphate dehydrogenase 1-like|KIAA0089GPD1-L|glycerol-3-phosphate dehydrogenase 1-like protein|EC 1.1.1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GPD1L (NP_055956.1, 1 a.a. ~ 351 a.a) full-length human protein.
- Purification:Protein G purified
- Cat. No.:103341-044
- Supplier no.:H00023171-D01P
Specifications
About this item
The GPD1L Antibody from Novus Biologicals is a rabbit polyclonal antibody to GPD1L. This antibody reacts with human. The GPD1L Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: GPD1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human