



Isopropylmagnesium chloride 2.0 M in THF, AcroSeal™
Catalog # 103524-182
Supplier: Avantor
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Colorless to yellow to grey to dark brown Solution (May develop some turbidity or precipitate)
- Titration Grignard:1.80 to 2.20 M
Specification Test Results
About this item
:102.84 g/mol
Boiling Pt:62.4 °C (899.8hPa)
Melting Pt:5 °C
Density:0.97 g/cm³
Flash Pt:–27 °C
Storage Temperature:Ambient
UN:3399
ADR:4.3,I
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:1068-55-9
:
Formula:C₃H₇ClMg
MDL Number:MFCD00000466