Order Entry
Puerto Rico
ContactUsLinkComponent
 
Plug Cap for Plain Top 15 ml Centrifuge Tubes, Globe Scientific
  10014-254
undefined
Plug Cap for Plain Top 15 ml Centrifuge Tubes, Globe Scientific
  10014-254
 :  6263C

 

  • Material:
    Polyethylene
  • Cat. No.:
    10014-254
  • Supplier no.:
    6263C

 

 

This cap is specifically designed for capping plain top 15 mL centrifuge tubes

Produced from polyethylene, this flanged plug cap provides exceptional leak-resistance.