Anti-Engrailed homeobox 2 Mouse Monoclonal Antibody [clone: 1E1.]
Catalog # 103325-728
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Engrailed homeobox 2
- Clonality:Monoclonal
- Clone:1E1.
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:2020
- Antigen Synonyms:engrailed-2|engrailed homolog 2|hu-En-2|Homeobox protein en-2|AUTS10|homeobox protein engrailed-2|engrailed homeobox 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:EN2 (NP_001418.2, 86 a.a. - 210 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GTCCAGAGGGRGGGAGGEGGASGAEGGGGAGGSEQLLGSGSREPRQNPPCAPGAGGPLPAAGSDSPGDGEGGSKTLSLHGGAKKGGDPGGPLDGSLKARGLGGGDLSVSSDSDSSQAGANLGAQP
- Purification:IgG purified
- Cat. No.:103325-728
- Supplier no.:H00002020-M03
Specifications
About this item
The engrailed homeobox 2 Antibody (1E1.) from Novus Biologicals is a mouse monoclonal antibody to engrailed homeobox 2. This antibody reacts with human. The engrailed homeobox 2 Antibody (1E1.) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Engrailed homeobox 2
Clonality: Monoclonal
Clone: 1E1.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human