To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GIT2
- Clonality:Monoclonal
- Clone:3B5-B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:9815
- Antigen Synonyms:MGC760|G protein-coupled receptor kinase interacting ArfGAP 2|G protein-coupled receptor kinase interactor 2|cool-associated|Cool-interacting tyrosine-phosphorylated protein 2|CAT-2|ARF GTPase-activating protein GIT2|tyrosine phosphorylated protein 2|ARF GAP GIT2|G protein-coupled receptor kinase-interactor 2|DKFZp686G01261|KIAA0148GRK-interacting protein 2|CAT2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GIT2 (AAH01379, 1 a.a. ~ 472 a.a) full length recombinant protein with GST tag.MSKRLRSSEVCADCSGPDPSWASVNRGTFLCDECCSVHRSLGRHISQVRHLKHTPWPPTLLQMVETLYNNGANSIWEHSLLDPASIMSGRRKANPQDKVHPNKAEFIRAKYQMLAFVHRLPCRDDDSVTAKDLSKQLHSSVRTGNLETCLRLLSLGAQANFFHPEKGNTPLHVASKAGQILQAELLAVYGADPGTQDSSGKTPVDYARQGGHHELAERLVEIQYELTDRLAFYLCGRKPDHKNGQHFIIPQMADS
- Purification:IgG purified
- Cat. No.:103337-640
- Supplier no.:H00009815-M01
Specifications
About this item
The GIT2 Antibody (3B5-B9) from Novus Biologicals is a mouse monoclonal antibody to GIT2. This antibody reacts with human. The GIT2 Antibody (3B5-B9) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: GIT2
Clonality: Monoclonal
Clone: 3B5-B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human