Anti-GIPR Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 591815]
Catalog # 103295-076
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:GIPR
- Clonality:Monoclonal
- Clone:591815
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μl
- Gene ID:2696
- Antigen Synonyms:MGC126722|GIP-R|gastric inhibitory polypeptide receptor|Glucose-dependent insulinotropic polypeptide receptor|PGQTL2
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:NS0 mouse myeloma cell line transfected with human GIPR (amino acids 1-466)
- Purification:Protein A or G purified
- Cat. No.:103295-076
- Supplier no.:MAB82103F
Specifications
About this item
The GIPR Antibody (591815) [FITC] from Novus Biologicals is a mouse monoclonal antibody to GIPR. This antibody reacts with human. The GIPR Antibody (591815) [FITC] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: GIPR
Clonality: Monoclonal
Clone: 591815
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human