Anti-FTS Rabbit Polyclonal Antibody
103284-008
- Antibody Type:Primary
- Antigen Symbol:FTS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:64400
- Antigen Synonyms:Fused toes protein homolog|fused toes homolog|FLJ13258|fused toes (mouse) homolog|FTSfused toes homolog (mouse)|Ft1|AKT-interacting protein|AKT interacting protein
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids: LNPEAAVLYEKDIQLFKSKVVDSVKVCTARLFDQPKIEDPYAISFSPWNP SVHDEAREKMLTQKKPEEQHNKSVHVAGLSWVKPGSVQPFSKEEK
- Purification:Immunogen affinity purified
- Cat. No.:103284-008
- Supplier no.:NBP2-14279
The FTS Antibody from Novus Biologicals is a rabbit polyclonal antibody to FTS. This antibody reacts with human, mouse, rat. The FTS Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: FTS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat