Anti-FosB/G0S3 Mouse Monoclonal Antibody (DyLight 350) [clone: 83B1138]
103359-814
- Antibody Type:Primary
- Antigen Symbol:FosB/G0S3
- Clonality:Monoclonal
- Clone:83B1138
- Conjugation:DyLight® 350
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:2354
- Antigen Synonyms:MGC42291|DKFZp686C0818|AP-1|GOSB|activator protein 1|G0S3G0/G1 switch regulatory protein 3|GOS3|FBJ murine osteosarcoma viral oncogene homolog B|oncogene FOS-B|protein fosB
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 2-15 of human FosB.
- Purification:Protein G purified
- Cat. No.:103359-814
The FosB / G0S3 Antibody (83B1138) [DyLight 350] from Novus Biologicals is a mouse monoclonal antibody to FosB / G0S3. This antibody reacts with human. The FosB / G0S3 Antibody (83B1138) [DyLight 350] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: FosB/G0S3
Clonality: Monoclonal
Clone: 83B1138
Conjugation: DyLight 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human