Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-FosB/G0S3 Mouse Monoclonal Antibody (DyLight 680) [clone: 83B1138]
Anti-FosB/G0S3 Mouse Monoclonal Antibody (DyLight 680) [clone: 83B1138]
  103359-798
Anti-FosB/G0S3 Mouse Monoclonal Antibody (DyLight 680) [clone: 83B1138]
  103359-798
 :  NB100-56530FR

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    FosB/G0S3
  • Clonality:
    Monoclonal
  • Clone:
    83B1138
  • Conjugation:
    DyLight® 680
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    2354
  • Antigen Synonyms:
    MGC42291|DKFZp686C0818|AP-1|GOSB|activator protein 1|G0S3G0/G1 switch regulatory protein 3|GOS3|FBJ murine osteosarcoma viral oncogene homolog B|oncogene FOS-B|protein fosB
  • Storage Buffer:
    50mM Sodium Borate
  • Storage Temperature:
    Store at 4C in the dark.
  • Immunogen:
    This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 2-15 of human FosB.
  • Purification:
    Protein G purified
  • Cat. No.:
    103359-798

 

 

The FosB / G0S3 Antibody (83B1138) [DyLight 680] from Novus Biologicals is a mouse monoclonal antibody to FosB / G0S3. This antibody reacts with human. The FosB / G0S3 Antibody (83B1138) [DyLight 680] has been validated for the following applications: Western Blot.

Type: Primary
Antigen: FosB/G0S3
Clonality: Monoclonal
Clone: 83B1138
Conjugation: DyLight 680
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human