To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:FMO2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:2327
- Antigen Synonyms:FMO 1B1|FLJ40826|Dimethylaniline oxidase 2|flavin containing monooxygenase 2 (non-functional)|flavin containing monooxygenase 2|dimethylaniline monooxygenase [N-oxide-forming] 2|pulmonary|Pulmonary flavin-containing monooxygenase 2|EC 1.14.13.8|FMO|FMO 2|FMO1B1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:FMO2 (NP_001451.1, 1 a.a. - 471 a.a.) full-length human protein. MAKKVAVIGAGVSGLISLKCCVDEGLEPTCFERTEDIGGVWRFKENVEDGRASIYQSVVTNTSKEMSCFSDFPMPEDFPNFLHNSKLLEYFRIFAKKFDLLKYIQFQTTVLSVRKCPDFSSSGQWKVVTQSNGKEQSAVFDAVMVCSGHHILPHIPLKSFPGMERFKGQYFHSRQYKHPDGFEGKRILVIGMGNSGSDIAVELSKNAAQVFISTRHGTWVMSRISEDGYPWDSVFHTRFRSMLRNVLPRTAVKWMIEQQMNRWFNHENYGLEPQNKYIMKEPVLNDDVPSRLLCGAIKVKSTVKELTETSAIFEDGTVEENIDVIIFATGYSFSFPFLEDSLVKVENNMVSLYKYIFPAHLDKSTLACIGLIQPLGSIFPTAELQARWVTRVFKGLCSLPSERTMMMDIIKRNEKRIDLFGESQSQTLQTNYVDYLDELALEIGAKPDFCSLLFKDPKLAVRLYFGPCNSY
- Purification:Protein A purified
- Cat. No.:103326-686
- Supplier no.:H00002327-D01P
Specifications
About this item
The FMO2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to FMO2. This antibody reacts with human. The FMO2 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: FMO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human