Anti-FGF R1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: M17D10]
Catalog # 103368-932
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:FGF R1
- Clonality:Monoclonal
- Clone:M17D10
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Size:100 μl
- Epitope:Epitope is within the sequence pro179 to lys195 in Ig loop II.
- Gene ID:2260
- Antigen Synonyms:FLGH3|BFGFR|EC 2.7.10|KAL 2|hydroxyaryl-protein kinase|EC 2.7.10.1|Proto-oncogene c-Fgr|heparin-binding growth factor receptor|HBGFR|KAL2|fms-related tyrosine kinase 2|CD331 antigen|FLT2H4|H5|CEK|Fms-like tyrosine kinase 2|OGD|fibroblast growth factor receptor 1|FLJ99988|FLT-2|CD331|soluble FGFR1 variant 2|H2|FGFR-1|bFGF-R-1|soluble FGFR1 variant 1|basic fibroblast growth factor receptor 1|FGFBR|N-SAM
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Recombinant human ectodomain of FGFr1b expressed in E. coli from pro23 to his415; antigen contained NH2-terminal gly-ser-pro-gly-ile and COOH-terminal glu-phe sequences.
- Purification:Protein G purified
- Cat. No.:103368-932
Specifications
About this item
The FGF R1 Antibody (M17D10) [PE] from Novus Biologicals is a mouse monoclonal antibody to FGF R1. This antibody reacts with human, mouse. The FGF R1 Antibody (M17D10) [PE] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: FGF R1
Clonality: Monoclonal
Clone: M17D10
Conjugation: PE (Phycoerythrin)
Epitope: Epitope is within the sequence pro179 to lys195 in Ig loop II.
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse