To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ERRFI1
- Antigen Symbol:ERRFI1
- Clonality:Monoclonal
- Clone:2B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:54206
- Antigen Synonyms:RALTreceptor-associated late transducer|ERBB receptor feedback inhibitor 1|GENE-33|MIG-6Mitogen-inducible gene 6 protein|MIG6
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ERRFI1 (NP_061821, 111 a.a. ~ 221 a.a) partial recombinant protein with GST tag.VVCGFKKLTVNGVCASTPPLTPIKNSPSLFPCAPLCERGSRPLPPLPISEALSLDDTDCEVEFLTSSDTDFLLEDSTLSDFKYDVPGRRSFRGCGQINYAYFDTPAVSAA
- Purification:IgG purified
- Cat. No.:103344-696
- Supplier no.:H00054206-M01
Specifications
About this item
The ERRFI1 Antibody (2B9) from Novus Biologicals is a mouse monoclonal antibody to ERRFI1. This antibody reacts with human. The ERRFI1 Antibody (2B9) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: ERRFI1
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human