Anti-Ephrin-B3 Rabbit Polyclonal Antibody
Catalog # 103260-708
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:Ephrin-B3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:1949
- Antigen Synonyms:ephrin-B3|EPH-related receptor tyrosine kinase ligand 8|EPLG8EPH-related receptor transmembrane ligand ELK-L3|Ephrin B3|LERK8EFL6|LERK-8
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:HSSPNYEFYKLYLVGGAQGRRCEAPPAPNLLLTCDRPDLDLRFTIKFQEYSPNLWGHEFRSHHDYYIIATSDGTREGLESLQGGVCLTRGMKVLLRVGQSPRGGAVPRKPVSEMPMERDRGAAHSLEPGKENLPGDPTSNATS
- Purification:Immunogen affinity purified
- Cat. No.:103260-708
- Supplier no.:NBP1-84829
Specifications
About this item
The Ephrin-B3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Ephrin-B3. This antibody reacts with human. The Ephrin-B3 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Ephrin-B3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human