Anti-EN1 Mouse Monoclonal Antibody [clone: 1F5]
Catalog # 103325-724
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Engrailed 1
- Antigen Symbol:EN1
- Clonality:Monoclonal
- Clone:1F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:2019
- Antigen Synonyms:Homeobox protein en-1|engrailed homeobox 1|hu-En-1|homeobox protein engrailed-1|engrailed homolog 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:EN1 (NP_001417, 266 a.a. - 392 a.a.) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SQQPLVWPAWVYCTRYSDRPSSGPRTRKLKKKKNEKEDKRPRTAFTAEQLQRLKAEFQANRYITEQRRQTLAQELSLNESQIKIWFQNKRAKIKKATGIKNGLALHLMAQGLYNHSTTTVQDKDESE*
- Purification:IgG purified
- Cat. No.:103325-724
- Supplier no.:H00002019-M06
Specifications
About this item
The EN1 / Engrailed 1 Antibody (1F5) from Novus Biologicals is a mouse monoclonal antibody to EN1 / Engrailed 1. This antibody reacts with human. The EN1 / Engrailed 1 Antibody (1F5) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: EN1
Clonality: Monoclonal
Clone: 1F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human