To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:DNMT3A
- Clonality:Monoclonal
- Clone:64B814.1
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:1788
- Antigen Synonyms:DNA cytosine methyltransferase 3A2|DNA (cytosine-5-)-methyltransferase 3 alpha|DNMT3A2|DNA (cytosine-5)-methyltransferase 3A|DNA MTase HsaIIIA|M.HsaIIIA|EC 2.1.1.37|Dnmt3a|DNA methyltransferase HsaIIIA
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:This antibody was raised against bacteria expressed HIS-tag recombinant mouse Dnmt3a.
- Purification:Protein G purified
- Cat. No.:103359-634
Specifications
About this item
The DNMT3A Antibody (64B814.1) [HRP] from Novus Biologicals is a mouse monoclonal antibody to DNMT3A. This antibody reacts with human, mouse. The DNMT3A Antibody (64B814.1) [HRP] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: DNMT3A
Clonality: Monoclonal
Clone: 64B814.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse